If you want to change the solvent type:
- To set the solvent type of A/B, run the script: A01 - Set Pump Solvent Types.
- To purge the system, run the script: B01 - Change Liquids / Solvents (with the following parameters: Solvents A/B and Refresh Only = Off).