The power supplies of the modules have wide-ranging capability, accepting any line voltage in the range specified for the modules.
CAUTION
Electric Shock or Damage to the Device
Connecting the device to a line voltage higher or lower than specified could result in personal injury or damage to the device.
The device can be affected by short voltage interruptions in case the power supply of the equipment is turned off to protect the device against an undetermined behavior. When the supply voltage returns, the power supply automatically restarts the device and reconnection to the chromatography data system is required.