The power cords are designed to match the wall socket requirements of the country in which they are used. The end of the power cords that plugs into the power socket on the device is identical for all power cords. The end of the power cords that plugs into the wall socket is different.
WARNING
Electric Shock or Damage to the Device
  • Never use a power cord other than the power cords provided by Thermo Fisher Scientific for the device.
  • Only use a power cord that is designed for the country in which you use the device.
  • Do not use extension cords.
  • Never plug the power cord to a power socket that is shared with other equipment (for example, multiple sockets).
  • Operate the device only from a power outlet that has a protective ground connection.
  • In case of emergency, it must be possible to reach the power cord easily at any time to disconnect the device from the power line.
WARNING
Electric Shock or Damage to a Product
Misuse of the power cords could cause personal injury or damage the instrument. Use the power cords provided by Thermo Fisher Scientific only for the purpose for which they are intended. Do not use them for any other purpose, for example, for connecting other instruments.