The Vanquish system is designed to be placed on a workbench. If the workbench is going to carry a complete Vanquish system and possibly other instruments, the bench must be capable of bearing the weight of all devices and instruments, including solvents.
For dimensions and weight, see Specifications.
Workbench Requirements
Part
Requirements
Site for the workbench
The workbench must stand in a secure and level position that is free of vibrations.
Workbench
  • Sturdy
  • Stable (non-tilting)
  • A height that ensures convenient access to the interior of each device in the system.
  • The bench top must be dry, clean, and resistant to chemicals.
  • If the system stack is to be stabilized with the Bench Clamp kit, the workbench top must fulfill one of the following requirements:
    • Thickness: Between 15 and 50 mm
    or
    • Material: Soft enough for wood screws
System Clearance Requirements
23145
Figure: System stack clearances (left: stack front view, right: stack top view)
No.
Description
1
Minimum 10 cm
2
Minimum 30 cm
3
Minimum 5 cm
4
Minimum 15 cm
5
Minimum 35 cm