Check the drain system for proper drainage of waste:
Pour water or isopropanol into the drain in the solvent rack.
Verify that the liquid leaves the system at the system drain port and into the waste container.
If a leak is visible, find and eliminate the problem.