Parts required
  • Drain tubing from ship kit
  • Elbow connection piece from ship kit
  • Suitable waste container
Follow these steps
  1. Push an elbow connection piece into the system drain port as far as it goes in to establish a self-sealing connection.
Connected system drain tubing and elbow connection piece
Figure: Connected system drain tubing and elbow connection piece
No.
Description
1
Funnel for system drainage
2
Detector waste outlet
2a
Detector waste line
3
Elbow connection piece
3a
System waste line
  1. Push the drain tubing into the elbow connection piece with your hand as far as it goes in to establish a self-sealing connection.
  1. Route the free end of the drain tubing into a waste container.
  1. Only if guiding the drain tubing toward the system's rear: Insert the drain tubing into the clips and make sure that the drain tubing does not form a siphon.
Inserting the drain tubing into clips
Figure: Inserting the drain tubing into clips
  1. Ensure proper drainage of waste (see next section).
Ensuring Proper Drainage of Waste
  1. Position the waste container below the level of the system stack. To prevent solvent vapors from evaporating back into the system, make sure that the distance is sufficient.
  2. Make sure the drain tubing is neither bent nor pinched.
  3. To prevent liquid from flowing back into the system, the entire drain tubing must remain below the drain port. Avoid the formation of siphons.
  4. To allow the waste liquid flowing off properly and avoid liquid accumulation in the system base, the end of the drain tubing must not be submerged in waste liquid (see figure below):
  1. Shorten the drain tubing if required.
  2. Observe the liquid level in the waste container and empty the waste container as necessary.
Free end of drain tubing in waste container
Figure: Free end of drain tubing in waste container