This section provides details on how to connect the modules to the power source. Do not turn on the system or modules yet.
NOTICE
Condensation in a device can damage the electronics.
  • Before connecting the devices to the power line, be sure that no condensation is present in the devices.
  • If you suspect that condensation is present, allow the device to warm up to room temperature slowly. Wait until the condensation is completely gone before proceeding.
Follow these steps
  1. Verify that the power switch on each device is set to OFF.
  2. Connect the power cord to the power-inlet connector on each device.
  3. Connect the free end of the power cords to an appropriate power source.
  4. Insert the signal and power cables into the cable clips (see Guiding Signal Cables in the Cable Clips).
  5. If necessary, move the system to its final position.
  6. Lock the system base against moving (see Locking the System Base).