When operating the system, make sure that the installation site meets these general environmental and operating conditions. For specified ambient temperature and humidity, see Specifications.
Temperature
Temperature fluctuations can affect the performance of the system. Avoid locations with significant changes in temperature and strong air drafts. For example, do not place the system in the direct sunlight, near heating or cooling sources, or under an air duct.
Humidity
The relative humidity of the operating environment is important for the performance of the system. Operate the system in the specified humidity range, with no condensation.
When the humidity is too high, condensation may occur, causing damage to the electronic components in the system. When the humidity is too low, static electricity may accumulate and discharge, shortening the life of the electronic components.
Ventilation
Make sure that the installation site is well ventilated at any time to avoid potential health hazards and safety risks, which may be caused by handling hazardous substances, volatiles or gases.
Vibration and mechanical shocks
Vibrations and mechanical shocks may affect the performance of the system. Therefore, the installation site should be free of vibrations. Avoid mechanical shocks and avoid placing the system in locations where vibrations are caused by other instruments.
Electromagnetic interference
  • The system has been tested and found to comply with the immunity requirements for both basic and industrial electromagnetic environments, pursuant to EN IEC 61326-1.
    Electromagnetic sources of strong interference may affect the performance of the system. In close proximity to the system, avoid operating equipment generating strong electric or magnetic fields and only operate certified laboratory equipment. Do not connect equipment that may cause power disturbances to the same power network used to supply the system.
  • The system has been tested and found to comply with the emission limits for a Class A equipment, pursuant to EN IEC 61326-1.
CAUTION
This equipment is not intended for use in residential environments and may not provide adequate protection to radio reception in such environments.