CAUTION
Possible Contamination
Hazardous substances may have contaminated the device during operation and may cause personal injury to service personnel.
Preparations
Remove the slide-in module that you want to return from the enclosure. See the related information:
Removing the Pump Slide-In Module
Follow these steps
Install the slide-in module to the enclosure of the replacement device. Follow the steps for inserting the slide-in module in the enclosure and tightening the captive screws in Installing the Slide-In Module.
If you need to return the device to Thermo Fisher Scientific for depot repair, contact your local Thermo Fisher Scientific support organization for the appropriate procedure.