CAUTION
Heavy Load, Bulky Device
The system modules are too heavy or bulky for one person alone to handle safely. To avoid personal injury or damage to the system modules, observe the following guidelines:
Tools required
Screwdriver, Torx T20
Preparations
When installing the slide-in module to an enclosure in the system stack, check that the enclosure is placed correctly in the stack.
Follow these steps
Push all tubing and capillaries, which are present in the tubing chase of the Vanquish Neo system modules, into the tubing chase. Otherwise, you will not be able to insert the slide-in module properly into the enclosure in the next step.
On the slide-in module, check that you can move the captive screws back and forth with your fingers. If you cannot, screw them in until you can.
Insert the slide-in module in the enclosure. The following steps require a team effort:
- Take the slide-in module on both sides from below.
- Lift the slide-in module to the height of the enclosure.
- Place the slide-in module in the enclosure so that the module sits in the enclosure by approximately 25 cm.
- Push the slide-in module onto the rails and into the enclosure until the slide-in module sits completely in the enclosure.
To facilitate tightening the slide-in module in the enclosure, pull out the module approximately 5 mm.
With the screwdriver, move each captive screw back and forth while pressing inward until the screw slips into the thread.
Push the slide-in module back into the enclosure as far as it goes in.
Gradually and evenly, tighten the four captive screws on the slide-in module hand-tight.
NOTICE
To set up the module, follow the steps in Installation.
Have a Thermo Fisher Scientific service engineer program the system serial number into the system module.