The system is designed to be operated mainly from a computer running with the chromatography software, and the Vanquish User Interface.
In addition, the following elements are available on the system:
Keypads:
The keypad buttons allow you to perform certain functions directly from the devices.
Status indicators:
Provide a quick visual check of the operational status of the device. The system provides the following status indicators:
Status indicator LED bar on the bottom on the front side of each device.
STATUS LEDs on the keypads.
System controller status LED