Capillaries
For detailed information on the usage of the capillaries, refer to the flow scheme from the ship kit.
Item
Quantity in shipment
Capillary, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x O.D. x length 20 µm x 360 µm x 550 mm, for nano LC applications
2
Capillary column-detector, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x O.D. x length 20 µm x 280 µm x 1 m, for nano LC applications
1
Capillary column-detector, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x O.D. x length 50 µm x 360 µm x 550 mm, for capillary and micro LC applications
4
Capillary pump-autosampler, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x O.D. x length 50 µm x 360 µm x 350 mm, for capillary and micro LC applications
1
Capillary autosampler-column/column compartment, nanoViper, fused silica-/PEEK-sheathed, 1/32", I.D. x O.D. x length 50 µm x 280 µm x 1 m, for capillary and micro LC applications
1
Capillary column-detector, fused silica, I.D. x O.D. x length 20 µm x 280 µm x 1.5 m, for nano LC applications
1
Capillary column-detector, fused silica, I.D. x O.D. x length 50 µm x 280 µm x 1.5 m, for capillary and micro LC applications
1
Capillary Accessories
Item
Quantity in shipment
Viper union
1
Fitting plug, Viper, titanium
3
Knurl for nanoViper fittings
2
Wash System Accessories
Item
Quantity in shipment
Tubing, silicone
3m
Tubing connector, straight (pack of 5)
1
Liquid Reservoirs and Liquid Reservoir Accessories
Item
Quantity in shipment
Bottle insert
1
Reservoir, 1 L, including cap
5
Reservoir, 0.25 L, including cap
4
Reservoir cap
10
Retaining guide (diameter 1.6 mm) to keep the following lines in the reservoir caps:
  • Solvent lines (reservoir to pump)
  • Needle wash liquid lines (reservoir to wash liquid degasser in pump and reservoir to autosampler wash pump)
10
Retaining guide (diameter 3.0 mm) to keep the seal wash line in the reservoir caps
10
Cap plug to close open holes in the reservoir cap (pack of 20)
2
Solvent line filter, filter holder (no filter frit or solvent line adapters included)
6
Solvent line adapter
6
Solvent line filter, filter frit, 10 μm
10
Tools
Item
Quantity in shipment
Seal-handling tool
1
Syringe, plastic, 12 mL
2
Torque screwdriver, Viper, base part with extension adapter and bits for tightening nanoViper capillaries and the autosampler needle seat
1
Tubing cutter for PEEK tubing
1
Tubing cutter for fused silica tubing
1
Cables
Item
Quantity in shipment
Digital I/O signaling cable, 5 m
1
LAN cable, (RJ45), 5 m
1
USB cable, type A to type B, high-speed, USB 2.0, length 0.5 m
To connect the pump to the autosampler.
1
USB cable, type A to type B, high-speed, USB 2.0, length 5 m
To connect the pump to the computer.
1
Autosampler-specific parts
Item
Quantity in shipment
Needle seat
1
Sample rack, for 54 x 12 mm O.D. vials
Note: The sample racks provide a 2D barcode for Vanquish rack type identification.
4
Sample container kit including well plates, mats, sealing tape, vials, and caps with pre-assembled septa
1
Miscellaneous
Item
Quantity in shipment
Drainage kit, Vanquish system, including:
  • L piece (elbow, pack of 4, different sizes)
  • Drain tubing, 11.4 m length
  • Clip (set of 2)
1
Fuses kit, Vanquish system
The kit includes the 5AT fuses for the pump, autosampler and the column compartment (optional).
1
Flow scheme
1
Cytochrome test sample
1
Paper label for magnetic label holder
1
Spiral tubing to wrap liquid lines or cables
1
Trap column cartridge holder
1
Acclaim™ PepMap™ 100 C18 HPLC column (set of 3)
1
Cap to close open ends of solvent lines (when disconnected from the pump)
6
Plug to close open connection ports in the pump (when no solvent lines are connected)
6