Flow connections between the modules of the Vanquish system are guided through either the tubing chase in the system or through guide holes or capillary clips.
Tubing Chase with Tubing Guides
To guide certain tubes and lines from the top module to the bottom module in the Vanquish Neo system stack, the stackable modules have a tubing chase on the inside right. The tubing chase provides four tubing guides.
Push the tubing or lines into the tubing guides.
Tubing chase with tubing guides (left: view from inside, right: view from top)
Figure: Tubing chase with tubing guides (left: view from inside, right: view from top)
Tubing Brackets
Tubing brackets are available for holding the tubing in place. Slip the bracket side onto the drain pipe.
Figure: Tubing bracket (left), tubing bracket installed (right)
Guide Holes and Capillary Clips
Guide holes and capillary clips are provided at specific positions on the system modules. Route flow connections from one module to the next module in the Vanquish Neo system through the appropriate guide hole or capillary clip when instructed to do so in the manual.