Allowed concentrations (standard system configuration):
System
(Standard Configuration)
Chloride
Buffer
Remarks
Vanquish Core
0.1 mol/L or less
1 mol/L or less
High chloride concentration: The application time should be as short as possible. Flush the system thoroughly after these applications.
Vanquish Horizon
Vanquish Flex
1 mol/L or less
-