To protect optimal functionality of the Vanquish system, observe these recommendations on the use of solvents and additives:
  • The system must be used with reversed-phase (RP) compatible solvents and additives only.
  • Use only solvents and additives that are compatible with all parts in the flow path.
TIP
In a Vanquish Core system, normal-phase (NP) compatible solvents and additives may be used if the system modules have been modified for NP applications. Refer to the Vanquish System Operating Manual.
Piston Seal Compatibility
  • In rare cases, a shortened lifetime of reversed-phase (UHMW-PE) piston seals has been observed with high pH, ammonium hydroxide containing mobile phases and prolonged exposure.