To restart the device after a long-term shutdown, follow these steps:
Turn on the device. Observe the following:
Situation | Action |
---|---|
If the device remained in the system stack and all system modules were turned off. | Turn on the system with the system power button on the system base. |
If the device is restarted after transport. | Turn on the device with the main power switch. |
Prepare and restart the other modules in the Vanquish system, following the instructions in the Operating Manuals for the modules. Pay special attention to the Preparing the Module for Operation section.
Before starting an analysis, let the device equilibrate and be sure that it is ready for operation. See Preparing the Device for Operation.