Cables and fuses
Description | Part No. |
---|---|
Fuses kit The kit includes the appropriate fuses for the Vanquish system modules. For the column compartment, use only 4 AT (5 x 20 mm), 250 V AC slow-blow fuses. | 6036.0002 |
USB cable, type A to type B, high-speed, USB 2.0 | 6720.8910A |
USB cable, type A to type B, high-speed, USB 2.0 | 6035.9035A |
Miscellaneous
Description | Part No. |
---|---|
Column compartment doors | 6732.3890 |
Column ID tags | 6732.0610 |
Column switching valve cover | 6732.1843 |
Immersion sensor PT100 For column compartment OQ/PQ | 6705.0060 |
Packing material | 6732.7004 |
PQ Kit For column compartment OQ/PQ, including a P700 thermometer and immersion sensor PT100. Note: To continue to use an already available PQ Kit 5705.0050A, it is required to order the temperature sensor clip. | 6732.0010 |
Rotor 6-p 7-p NP VC-C | 6230.2537 |
Stator 6-p 7-p NP VC-C | 6230.2535 |
Temperature sensor clip For column compartment OQ/PQ | 6732.0009 |
Viper inline filter To retain particles (larger than 0.5 µm) from the mobile phase | 6036.1045 |
Viper union | 6040.2304 |
For active pre-heaters, passive pre-heaters and post-column coolers, see Optional Accessories. |
Power cords
Description | Part No. |
---|---|
Power cord, Australia | 6000.1060 |
Power cord, China | 6000.1080 |
Power cord, Denmark | 6000.1070 |
Power cord, EU | 6000.1000 |
Power cord, India, SA | 6000.1090 |
Power cord, Italy | 6000.1040 |
Power cord, Japan | 6000.1050 |
Power cord, UK | 6000.1020 |
Power cord, USA | 6000.1001 |
Power cord, Switzerland | 6000.1030 |