Item | Part No. |
---|---|
Active pre-heater, 0.1 x 380 mm, MP35N | 6732.0110 |
Active pre-heater, 0.1 x 610 mm, MP35N | 6732.0150 |
Conversion kit For the extended configuration of up to three column compartments, more than one conversion kit may be required. Only service personnel certified by Thermo Fisher Scientific are permitted to install the extended configuration with up to three column compartments or to perform the left-side installation of the column compartment. | Contact Sales |
Passive pre-heater, 0.1 mm x 530 mm, MP35N (VH-C) | 6732.0174 |
Passive pre-heater, 0.1 mm x 680 mm, MP35N (VH-C) | 6732.0184 |
Passive pre-heater, 0.18 mm x 530 mm, stainless steel (VC-C) | 6732.0170 |
Post-column cooler, 1µL, 0.1 x 240 mm (VH-C10 only) | 6732.0510 |
Post-column cooler, 1µL, 0.1 x 590 mm (VH-C10 only) | 6732.0520 |
Valve 2-p 6-p VC-C10 The column compartment can hold one or two column switching valves. | 6230.1520 |
Valve 6-p 7-p VC-C10 The column compartment can hold one or two column switching valves. | 6230.1530 |
Valve 2-p 6-p 150MPa bio VH-C/VC-C The column compartment can hold one or two column switching valves. | 6036.2520 |
Valve 6-p 7-p 150MPa bio VH-C/VC-C The column compartment can hold one or two column switching valves. | 6036.2530 |
Valve 6-p 7-p 70 MPa NP VC-C | 6230.2530 |