The device performance is specified as follows:
Type
Specification
Capacity
2 column slots
Pressure range
VC-C10: 2 – 70 MPa (20 – 700 bar, 290 – 10100 psi)
VH-C10: 5 – 151 MPa (50 – 1517 bar, 700 – 22002 psi)
Temperature range (column chamber)
VC‑C10: 5 °C – 85 °C, max. 18 °C below ambient
VH‑C10: 5 °C – 120 °C, max. 18 °C below ambient
Temperature accuracy (column chamber)
±0.5 °C (up to 80 °C)
Temperature stability (column chamber)
±0.05 °C
Temperature precision (column chamber)
±0.1 °C
Heat up time (column chamber)
from 20 °C to 50 °C in less than 15 minutes ±1 °C
from 25 °C to 40 °C in 5 minutes (typically) ±1 °C
Cool down time (column chamber)
from 50 °C to 20 °C in less than 15 minutes ±1 °C
Column switching valves
2-position-6-port valve
6-position-7-port multi-position valve
Communication
1 USB port (USB 2.0, "B" type connector)
System Interlink
2 system interlink ports (RJ45-8 connectors)
Control
Chromeleon 7
The device can be operated also with other data systems. For details, contact the Thermo Fisher Scientific sales organization.
Keypad with 4 buttons for performing certain functions directly from the device.
Materials in the flow path
Valve: ceramic/titanium; capillaries: MP35N or stainless steel (for details, see the pre-heater and post-column cooler specifications)
NOTICE For information about the chemical resistance of materials refer to the technical literature.
Solvent and additive information
See Solvent and Additive Information.
Safety features
Liquid leak sensor
Good Laboratory Practice (GLP) features
Predictive Performance functions for scheduling maintenance procedures based on the actual operating and usage conditions of the device.
All system parameters logged in the Chromeleon Audit Trail.
Active pre-heaters
Type
Specification
Temperature range
40 °C to upper temperature limit of column chamber
5 °C above environment and / or eluent temperature at active pre-heater input
Wetted parts
MP35N
Biocompatible
Yes
Heating performance
80 °C at 2.5 mL/min
120 °C at 1 mL/min
Temperature accuracy
± 2 °C (up to 80 °C)
Temperature stability
± 0.5 °C
Temperature precision
± 1 °C
Heated volume
0.9 µL
Total volume
0.1 x 380 mm:
3 µL
0.1 x 610 mm:
6 µL
Passive pre-heaters
Type
0.18 x 530 mm, stainless steel
0.1 x 530 mm, MP35N
0.1 x 680 mm, MP35N
Temperature range
Same as column chamber
Same as column chamber
Same as column chamber
Biocompatible
No
Yes
Yes
Heated volume
3 µL
1 µL
1 µL
Total volume
15 µL
5 µL
6 µL
Post-column coolers (VH-C10 only)
Type
Specification
Temperature range
40 °C to 80 °C
Wetted parts
MP35N
Biocompatible
Yes
Cooling performance
down to 50 °C at 2 mL/min and 120 °C compartment temperature
Temperature accuracy
± 2 °C (up to 80 °C)
Temperature stability
± 0.5 °C
Temperature precision
± 1 °C
Cooled volume
1 µL
Total volume
0.1 x 240 mm:
3 µL
0.1 x 590 mm:
6 µL