To prepare the device for transport, follow these steps:
Perform a long-term shut down of the device. See Long-Term Shutdown.
Turn off the device with its main power switch and disconnect the power cord.
Remove all cables and flow connections to other devices.
Cover up the valve ports with the fitting plugs.
Remove the post-column cooler (if installed). For details, see Replacing the Post-Column Cooler (VH-C10 only).
Remove the pre-heater, if installed:
For removing the active pre-heater, see Replacing the Active Pre-Heater.
For removing the passive pre-heater, see Replacing the Passive Pre-Heater.
Remove the column:
- Disconnect the capillaries from the column inlet and outlet.
- Detach the column holder from the central wall inside the column chamber.
- Detach the column holder from the column fitting.
Remove the column compartment from the Vanquish system:
Grasp the column compartment by its sides and slightly lift the column compartment up and out of the grooves on the mounting bars.