When
Column compartment door is defective
Parts required
Column compartment door
Preparations
Open the column chamber cover. See Accessing the Interior Components.
Follow these steps
  1. Open the door to an angle of 90°, a position in which the two hinges on the housing are in the grooves on the door.
  2. To release the door, push it upward.
  3. Slightly tilt the door to the outside, away from the housing, and remove the door.
Hinging the door
Figure: Hinging the door
No.
Description
1
Hinge on the housing
2
Groove on the door
3
Pin
  1. To install the door, align the grooves on the door with the hinges on the housing.
  2. Insert the lower hinge and pin in the groove on the door.
  3. Slightly push up the door to insert the upper hinge and pin.
  4. Push the door downward to lock it in place.
    You can close the door only when it is properly installed.