Parts required
Column switching valve cover
Tools required
Screwdriver, Torx T10
Removing the Column Switching Valve Cover
  1. Open the column compartment door.
  2. With the screwdriver, loosen the 4 screws on the switching valve cover.
  3. Figure: Switching valve cover screws
  4. Remove the screws and keep them for further installation.
  5. Remove the switching valve cover and gasket.
  6. 12
    Figure: Removing the switching valve cover and gasket
    No.
    Description
    1
    Gasket
    2
    Cover
  7. Carefully detach the cable from the rear of the switching valve gasket.
Installing the Column Switching Valve Cover
  1. Attach the cable to the rear of the new column switching valve gasket/cover.
  2. Position the gasket and the switching valve cover onto the column switching valve opening.
  3. With the screwdriver, tighten the 4 screws on the column switching valve cover.