The figure shows the recommended fluidic setup for the use of one column switching valve.
Example: Connections on a 2-p, 6-p column switching valve, upper valve setup, left: position 6-1, right: position 1-2
Figure: Example: Connections on a 2-p, 6-p column switching valve, upper valve setup, left: position 6-1, right: position 1-2
Follow these steps
  1. Connect the Viper capillary from the pump or autosampler to port 6 of the column switching valve.
  2. Depending on the column position, connect an active or passive pre-heater to port 1 or 5 of the column switching valve and connect it to the column inlet.
  3. Connect a Viper capillary to port 2 or 4 of the column switching valve and connect it to the column outlet.
  4. Connect a post-column cooler or a Viper capillary to port 3 of the column switching valve and connect it to flow cell inlet of the detector.