This section describes how to connect and place the active pre-heater in the column compartment.
Parts required
  • Column holder
  • Active pre-heater
  • Viper inline filter (optional)
Preparations
NOTICE
The active pre-heater is filled with isopropanol when the column compartment is shipped from the manufacturing site. If of insufficient quality, isopropanol may damage the column.
  • Flush the components filled with isopropanol to waste, using solvents that are miscible with isopropanol. If they are not, use an appropriate solvent of similar polarity.
  • Install a Viper union in the flow path that replaces the column for the flushing procedure.
Follow these steps
TIP
Use a Viper inline filter between the active pre-heater and the column to prevent un-dissolved particles from getting into the column.
  1. Connect the Viper inline filter to the column.
  2. Connect the active pre-heater to the Viper inline filter.
  3. Attach the column holder to the active pre-heater fitting.
  4. Fix the column holder on the central wall inside the column chamber.
  5. Positioning pre-heater and column inside the column chamber
    Figure: Positioning pre-heater and column inside the column chamber
  6. Before you connect the pre-heater cable to the pre-heater connection port for the first time, check that the pre-heater is enabled in the Instrument Configuration Manager dialog box for the column compartment.
    Once the pre-heater is enabled in the chromatography data system, it detects automatically when the pre-heater is connected.
  7. Connect the pre-heater cable to the pre-heater connection port.
    The pre-heater connection ports are located next to the lower column switching valve.
  8. TIP
    Use the left connection port for a pre-heater installed in the left column slot, and the right connection port for a pre-heater installed in the right column slot.
  9. Pre-heater connection port
    Figure: Pre-heater connection port
  10. Route the pre-heater cable through the pre-heater cable guide. Each pre-heater cable and cable guide is located above the pre-heater port. If the pre-heater cable is too long, loop the cable in the guide.
12
Figure: Pre-heater cable and cable guide; on the right: cable looped in guide
No.
Description
1
Cable guide
2
Pre-heater cable