Parts required
  • Drain tubing
  • L-piece
  • Waste container as required
  • Tubing cutter (optional)
Follow these steps
  1. Connect the drain tubing to the L-piece.
  2. Connect the L-piece to the drain port.
    The drain port is located on the bottom front of the device, below the front door.
  3. Connecting drain tubing
    Figure: Connecting drain tubing
    No.
    Description
    1
    Drain port
    2
    L-piece
  4. Place the free end of the drain tubing into an appropriate waste container.
    If you should ever have to cut tubing to length, use a tubing cutter. Make sure that the cut is at a right angle to the length of the tubing and that the end is free of nicks or burrs.
NOTICE
To maintain a positive siphon, position the waste container below the level of the column compartment. Otherwise, liquid may flow back into the interior and cause damage to the device.
For information about how the liquid is discharged to waste through the Vanquish drain system, refer to the Vanquish System Operating Manual.