TIP
Before you attach the column compartment to the system, connect the waste line to the system drain port. For instructions, refer to the Vanquish System Operating Manual.
Parts required
  • 4 T-slot nuts
  • 2 crosspieces
  • 4 screws
  • 4 spacers
Tools required
Screwdriver, Torx T20
Follow these steps
  1. Align the screw holes of the two T-slot nuts, the spacers and the crosspiece.
  2. Mounting bar
    Figure: Mounting bar
    No.
    Description
    1
    Spacer
    2
    Groove
    3
    Screw
    4
    T-slot nut
    5
    Crosspiece
  3. With the screws, fix the T-slot nuts and the spacers onto the crosspiece.
    The T-slot nuts must face each other, as shown above.
  4. Slide the T-slot nuts of the mounting bar in the rail on the system housing and push until stop.
    The grooves on the mounting bars must face upwards. Make sure, that the orientation of the crosspiece is as indicated on the picture.
  5. Tighten the screws on the mounting bars.
  6. Installing the mounting bars on the system housing
    Figure: Installing the mounting bars on the system housing
    No.
    Description
    1
    Rail
    2
    Mounting bar
  7. Lift the column compartment into vertical position.
  8. To insert the holding knobs in the grooves on the mounting bar, grasp the column compartment by its sides and slightly lift the column compartment up.
Attaching the column compartment to the system stack
Figure: Attaching the column compartment to the system stack