The user-accessible components of the column compartment are located behind the front door:
1235789101234566789104
Figure: Interior view (left: with chamber cover; right: without chamber cover)
No.
Description
1
Connection ports for column ID tags
2
Upper column switching valve (if installed)
3
VH-C10 only: Post-column cooler (if installed)
4
Capillary guides
5
Column chamber (with and without chamber cover)
6
Passive pre-heater mounting positions
7
Connection ports for active pre-heater
8
Lower column switching valve (if installed)
9
Leak tray with sensor for liquid leaks
10
Keypad with four buttons for device control