When
The leak sensor is wet. The leak sensor reports leakage.
Parts and additional items required
  • Replacement part as required
  • Cloth or tissue
Preparations
When resolving leakage, observe the safety guidelines and general rules for maintenance and service as presented in Maintenance and Service.
Follow these steps
  1. Locate the source of the leak.
    As leakage usually occurs at a connection, visually inspect all components and connections in the flow path.
  2. Tighten or replace the connection or component as required.
  1. With a cloth or tissue, thoroughly absorb all liquid that has collected in the leak tray and under the leak sensor. Be careful not to bend the sensor.
  2. Allow the sensor to adjust to the ambient temperature for a few minutes.
  3. If leakage is no longer reported, you can resume operation.