The Chromeleon software supports functions for monitoring and recording service and qualification information about the device.
Monitoring service and qualification intervals
On special service and qualification panels, you can define intervals for service procedures or qualification procedures. These functions, which are called Predictive Performance, allow you to schedule these procedures based on the actual operating and usage conditions of the device. In addition, you can set limits to alert you before and when the service or qualification is due.
Color-coded bars provide visual feedback, allowing you to easily check and monitor the status. If a warning limit was set, a message in the Chromeleon Audit Trail alerts you when the action is due.
Service and qualification counters can be reset to zero after the required action was performed. To keep the Predictive Performance information up-to-date, consider resetting the counter when a service, or qualification procedure has been performed.
For more information, refer to the Chromeleon Help.