Pay attention to the following safety guidelines:
Observe all warning messages and precautionary statements presented in Safety Precautions.
CAUTION
Tip-Over
This column compartment can tip over.
CAUTION
Electric Shock or Damage to the Device
After the power to the device is turned off, the device is still energized as long as the power cord is connected. Repair work on the device while the device is connected to power could lead to personal injury.
NOTICE
To lift or move the device, grasp the device by the sides. Do not move or lift the device by the front door. This will damage the door or the device.