The physical conditions of the device are specified as follows:
Type | Specification |
---|---|
Range of use | Indoor use only |
Ambient operating temperature | 5 °C - 35 °C |
Ambient storage temperature | -20 °C - 45 °C |
Ambient operating humidity | 20% - 80% relative humidity (non-condensing) |
Ambient storage humidity | Maximum 60% relative humidity (non-condensing) |
Operating altitude | Maximum 2000 m above sea level |
Pollution degree | 2 |
Power requirements | 100 – 240 V AC, ± 10 %; 50/60 Hz, max. 310 W / 330 VA |
Overvoltage category | II |
Emission sound pressure level | < 50 dB(A), typically < 45 dB(A) |
Dimensions | 70 x 11.1 x 42 cm |
Weight | 13 kg, no column switching valves installed, 14 kg, two column switching valves installed |