The status LED bar on the front side of the device and the STATUS LED on the inside keypad provide information about the device status.
LED Bar
The LED bar colors provide the following information:
LED Bar
Description
Off (dark)
The power to the device is turned off.
Dimmed
The door of the device is open.
Yellow, flashing slowly
The power to the device is turned on, but the device is not connected in the chromatography data system.
Yellow
The device is connected in the chromatography data system.
Green, flashing
The device is equilibrating.
Green
The device is equilibrated.
Blue
An injection or sequence is running, including data acquisition.
Red
A problem or error has occurred. For the related message, check the Instrument Audit Trail. For remedial action, see Troubleshooting.
STATUS LED
The STATUS LED on the keypad inside the device provides the following information:
STATUS LED
Description
Off (dark)
The power to the device is turned off.
Green
The device is functioning properly.
Red
A problem or error has occurred. For the related message, check the Instrument Audit Trail. For remedial action, see Troubleshooting.