The device is designed to be operated mainly from a computer running with the chromatography software.
In addition, the following elements are available on the device:
  • Keypad
    The keypad buttons allow you to perform certain functions directly from the device.
  • Status indicators
    The LEDs (Light Emitting Diodes) on the status indicator LED bar on the front side of the device and the STATUS LED on the keypad provide a quick visual check of the operational status of the device.