System Arrangement - Column Compartment H - Column Compartment C - Vanquish Column Compartments (VC-C10, VH-C10) Operating Manual - Installation - System Arrangement - Setting Up the Hardware - Column Compartment H - Column Compartment C
Chromatography > Liquid Chromatography > Vanquish Series > Vanquish Horizon UHPLC > Column Compartment H
Chromatography > Liquid Chromatography > Vanquish Series > Vanquish Core HPLC > Column Compartment C
Document Type
Operating Manual
The device is part of the Vanquish system. The system modules are typically arranged in a system stack, with the arrangement depending on the system configuration.
Figure: Vanquish system, standard configuration (example)
No.
Description
1
Solvent Rack
2
Detector
3
Autosampler
4
Pump
5
System Base
6
Column Compartment
In the standard configuration, the Vanquish system includes one column compartment, installed to the right side of the system.
For instructions on how to set up the system stack, refer to the Vanquish System Operating Manual.
TIP
An extended or left-side configuration of the column compartment is also available. Observe the following notes:
The extended configuration on the right side can include up to three column compartments. For the extended configuration of the column compartment, up to two conversion kits are required.
The column compartment can also be installed to the left side of the Vanquish system stack. For the left-side installation, one column compartment requires one conversion kit. Each additional column compartment on the left side requires an additional conversion kit. If a Vanquish Charger or the Vanquish Display is used with the Vanquish system, the column compartment must be attached to the right side of the system.
For ordering information, contact your local Thermo Fisher Scientific sales organization.
Installation by Service Engineer only
Only service personnel certified by Thermo Fisher Scientific are permitted to install the extended configuration with up to three column compartments or to perform the left-side installation of the column compartment.