WARNING
Electric Shock or Damage to the Device
High voltages are present inside the device that could cause an electric shock or damage to the device.
  • Do not make any changes to the electrical or grounding connections.
  • If you suspect any kind of electrical damage, disconnect the power cord and contact Thermo Fisher Scientific Technical Support for assistance.
  • Do not open the housing or remove protective panels unless specifically instructed to do so in this manual.
  • Do not place liquid reservoirs directly upon the device. Liquid might leak into the device and get into contact with electronic components causing a short circuit. Instead, place liquid reservoirs in the solvent rack that is available for the Vanquish system.