Allowed concentrations for the charged aerosol detector:
Concentration | Value |
---|---|
Volatile buffer concentration | Less than or equal to 0.1 mol/L |
Chloride concentration | Not recommended |
Observe the mobile phase guidelines for the detector. See Mobile Phase Guidelines for the Detector.