CAUTION
Escaping Gas
Gas can escape from open gas connections when the detector gas flow and/or the gas supply is turned on. The escaping gas can pose a health and safety risk.
Parts and tools required
Only required if gas tubing is to be replaced
For replacement of the gas inlet tubing:
Gas inlet elbow connector
Gas inlet tubing
For replacement of the gas exhaust tubing:
Gas exhaust elbow connector
Gas exhaust tubing
Tubing cutter (optional)
Preparations
Flush the detector with the detector gas flow being turned on. For recommendations on flushing the detector, see Flushing the Detector Between Analyses.
Turn off the pump flow and then the gas flow:
- Turn off the pump flow to the detector. Wait at least 5 minutes. During the waiting period, the detector drains any remaining mobile phase to waste.
- Turn off the gas flow on the detector.
- Wait until the system pressure is down to zero before you continue.
- Turn off the gas supply to the detector.
Turn off the detector with the main power switch.
Follow these steps
On the gas port on the detector, disconnect the elbow connectors of the gas tubes from the port.
To re-install the gas tubes or install new gas tubes, follow the steps in Connecting the Gas Inlet and Exhaust Tubes.