The following signal channels are available for the detector:
CAD_1
The signal channel is available for data acquisition.
Background current
The signal channel displays the background current of the detector during data acquisition without autozero or filter constant applied.
It is recommended to always record the background current signal channel.
If a problem occurs, the information from the signal channel can provide helpful information to identify and eliminate the source for the problem.
Gas regulator pressure
The signal channel records and monitors the nebulizer gas pressure during data acquisition.
It is recommended to record the signal channel for troubleshooting purposes.
For proper gas pressure regulation, or if baseline noise or poor precision occurs, the information from the signal channel can provide helpful information to identify and eliminate the source for the problem.
Evaporation tube temperature
The signal channel records and monitors the actual temperature of the evaporation tube during data acquisition.
It is recommended to always record the evaporation temperature signal channel for troubleshooting purposes.
If a problem occurs, the information from the signal channel can provide helpful information to identify and eliminate the source for the problem.