The following table serves as an overview of parameters that influence the performance of the detector during data acquisition.
Parameter
Affects
Filter constant
Sensitivity, baseline noise
Evaporation temperature
Baseline noise, background current
Power function value
Linear range
Data collection rate
Peak resolution, disk space, possibly baseline noise
For further information about the parameters, refer to the Chromeleon Help.