Before operating the detector, verify the nebulizer gas pressure setting and the nebulizer serial number.
NOTICE
Operating the nebulizer with a gas pressure other than the pressure specified for this nebulizer can cause unpredictable deviations in performance of the nebulizer and the detector. After installation of the nebulizer, set the nebulizer gas pressure to the pressure that is specified for your nebulizer. The nebulizer gas pressure is defined on the gas pressure certificate that is shipped with the nebulizer.
  1. In Chromeleon, open the Command window. Navigate to the Gas Regulator Pressure Nominal parameter.
  2. Check if the gas pressure on the nebulizer certificate matches the nebulizer gas pressure set in the Gas Regulator Pressure Nominal parameter.
    If the nebulizer gas pressure deviates from the pressure on the nebulizer certificate, enter the gas pressure from the nebulizer certificate.
  3. Check whether the nebulizer serial number on the nebulizer certificate matches the nebulizer serial number set in the Nebulizer Serial Number parameter.
    If the nebulizer serial number deviates from the serial number on the nebulizer certificate, enter the serial number from the nebulizer certificate.
  4. Close the Command window.
  5. Monitor the Gas Regulator Pressure until it reaches the setpoint.