Minimizing non-volatile matter in the mobile phase
NOTICE
Long-term exposure of the detector to non-volatile mobile phases will damage the detector. Do not use non-volatile buffers.
Observe the following guidelines to minimize the concentration of non-volatile matter in the mobile phase:
  • Prepare all mobile phases with volatile solvents. As a general rule, use solvents that are at least as volatile as water, that is lower boiling point and higher vapor pressure.
  • Use ultra-pure water with high resistivity and low total organic carbon obtained fresh from a thoroughly flushed and well-maintained point-of-use purification system.
  • TIP
    Bottled water and stagnant water from a purification system typically contain a higher level of non-volatile impurities than freshly obtained ultra-pure water.
  • Choose solvents that are specified to have low residue after evaporation.
    For example, LC/MS-grade solvents typically have a lower residue after evaporation specification than HPLC-grade solvents and are often the better choice to achieve consistently low baseline noise.
  • TIP
    Many organic solvents have low viscosity, low surface tension and high volatility, which lead to a high mass transport efficiency within the detector. For such solvents, it is especially recommended to select a grade with lowest residue after evaporation.
  • If a particular solvent causes high background current or noise, try a different grade or vendor.
  • Most HPLC-grade or better solvents are pre-filtered and additional filtration may increase the concentration of impurities within the mobile phase. It is therefore recommended to only filter the aqueous buffer salt portion of a mobile phase.
  • TIP
    With solvent gradients, non-volatile impurities in the more polar solvent (for example, water in reversed-phase) can be concentrated on the column and elute later to produce large peaks or baseline artifacts.
  • With highly combustible mobile phases, such as tetrahydrofuran (THF) and other ethers or ketones, use only nitrogen as supply gas.
  • When changing the operating mobile phase: Make sure that the solvents are miscible and will not cause precipitation of buffer salts. If required, use an appropriate intermediate solvent.
  • TIP
    Keep this in mind particularly when switching between chromatographic methods.
  • Water or highly aqueous (< 5% organic modifier) mobile phases are highly susceptible to microbial growth. Prepare such mobile phases freshly on a daily basis.
  • Select an optimal evaporation temperature to selectively evaporate background components of the mobile phase. Observe the guidelines in Evaporation Temperature.
  • If the detector is inadvertently exposed to a non-volatile mobile phase, use water or appropriate solvent with no additives to remove the salts from the detector.
Typical compatible solvents for use with the detector
  • Water
  • Methanol
  • Isopropanol
  • Acetonitrile
  • Acetone
  • Methylene chloride
  • Hexane
  • Chloroform
  • Tetrahydrofuran
    NOTE: Use THF with nitrogen gas only.