Observe the guidelines in the following sections for mobile phase use with the charged aerosol detector:
CAUTION
Explosion Hazard or Damage to the Detector
When using tetrahydrofuran (THF), the use of air as supplied gas may pose an explosion hazard. This can pose a safety and health risk and damage the detector.
NOTICE
Inappropriate or contaminated mobile phase can damage the detector. The quality of the mobile phase significantly influences the performance of the detector. Observe the following guidelines to avoid damage to the detector.