If the charged aerosol detector is the only detector in the Vanquish system flow path, connect the inlet capillary between column and nebulizer.
Preparations
See Connecting the Inlet Capillary to the Nebulizer.
Follow these steps
  1. Connect the capillary to the column outlet.
  2. Route the inlet capillary from the column compartment through the guide hole in the detector enclosure to the nebulizer. Use the guide hole that is next to the column compartment.
  3. Connect the inlet capillary to the nebulizer.
Inlet capillary and waste line connected to/from the detector
Figure: Inlet capillary and waste line connected to/from the detector