NOTICE
Extending or combining the waste line can cause a backpressure in the line that can lead to an excess liquid build-up inside the detector. Avoid extending or combining the waste line. Make sure that the waste line drains freely into waste.
Parts required
Waste line for connection between waste outlet and waste container
Tools required
Tubing cutter (optional)
Preparations
  1. Route the waste line through the detector waste outlet in the system base to waste. Follow the instructions in the Vanquish System Operating Manual.
  2. On the waste outlet on the detector, remove the plug.
Follow these steps
  1. Route the waste line from the system base to the detector, through the tubing guides of the system modules below the detector.
  2. On the waste outlet on the detector, connect the waste line.
  3. Waste line connected to the waste port
    Figure: Waste line connected to the waste port
    No.
    Description
    1
    Waste outlet
    2
    Waste line
  4. Check the waste line over the entire flow path. Make sure that no bending (kink), pinching or squeezing of the waste line is present at any point in the flow path.
  5. If you have to cut tubing to length, use a tubing cutter. Make sure that the cut is at right angle to the length of the line and that the cut is not crimped.
TIP
The waste line should go straight to the system base and to waste. Make sure that the line is positioned straight in the tubing guides.