CAUTION
Explosion Hazard or Damage to the Detector
When using tetrahydrofuran (THF), the use of air as supplied gas may pose an explosion hazard. This can pose a safety and health risk and damage the detector.
CAUTION
Excessive Gas Supply Pressure
An improper or excessive gas supply pressure can lead to an overpressure in the detector that can cause the safety valve in the detector to release gas. In extreme cases, the excessive pressure can cause the gas filters to rupture. This can pose a health and safety risk.
CAUTION
Gas Filter Rupture upon Excessive Gas Supply Pressure
An excessive supply gas pressure can lead to an overpressure in the detector and can cause the gas filters to rupture. This can pose a health and safety risk.
NOTICE
Gas Specifications
The requirements and specifications for the gas supplied to the detector have major impact on the detector performance. Observe the following notes:
Make sure that the location fulfils the following requirements for the gas supply:
The gas supply and the gas supply pressure for the detector must meet the gas specifications. See Performance Specifications.
Make sure that the gas supply is regulated to a stable gas pressure that is within the specified gas pressure range.
The gas supplied to the detector must be either air or nitrogen. Do not use helium or other supply gases.
Nitrogen gas (typically ≥ 95% nitrogen purity) is recommended for most applications. Air may be used when working with mobile phases of low combustibility.
With highly combustible mobile phases, such as tetrahydrofuran (THF) and other ethers or ketones, use only nitrogen as supply gas.
The use of a non-bottled gas source is recommended, such as
A nitrogen generator with suitable air compressor or with a suitable in-house air source
Medical-grade nitrogen from an in-house liquid nitrogen source
TIP
Use of smaller compressed nitrogen cylinders or liquid nitrogen containers may be a quick and easy source of nitrogen during installation. However, it is not recommended for routine operation.
The gas consumption of the detector is typically ≤ 4 L/min.
The supply gas must be free from water vapor, free from particles, and must not contain non-volatile hydrocarbons, such as compressor oils.
If necessary, use a sub-micron particle filter or carbon filter in combination with a water condensation trap located close to the gas source.
Make sure that the gas supply is ready for operation. Follow the instructions in the User Documentation for the gas supply.