The FocusJet nebulizer is the liquid flow inlet of the charged aerosol detector. Eluate from the column flows into the nebulizer and merges with filtered gas that is supplied to the nebulizer. Due to the concentric flow design of the nebulizer, an aerosol is produced within the spray chamber.
FocusJet nebulizer
Figure: FocusJet nebulizer