<Overview_InteriorComp>The user-accessible components of the detector are located directly behind the front doors:
Interior view
Figure: Interior view
No.
Description
1
Keypad with status indicators
2
Gas filter assembly behind the protective cover
3
Leak tray
4
Nebulizer (after installation)
5
Waste outlet
6
Type label, indicating the module name, serial number, part number, and revision number (if any)
7
Drain port
8
Leak sensor