The power supply of the device has wide-ranging capability, accepting any line voltage in the range specified for the device.
To avoid power fluctuations, the use of an uninterruptible power supply (UPS) is recommended.
CAUTION
Electric Shock or Damage to the Device
Connecting the device to a line voltage higher or lower than specified could result in personal injury or damage to the device.
Connect the device to the specified line voltage only.