Device Connectors
The following connectors are provided on the device:
Electrical and gas connectors on the right side of the detector
Figure: Electrical and gas connectors on the right side of the detector
No.
Description
1
Main power switch (on/off control)
2
Fuse holder
3
Power-inlet connector
4
System Interlink port
Allows power on/off control for the detector from the Vanquish system base and device communication
For example, the interconnection between charged aerosol detector and pump enables direct communication in case of an operating issue. Certain problems in the charged aerosol detector cause an automatic stop of the pump flow to prevent a build-up of excess liquid inside the detector.
5
USB hub ("A" type connector)
Allows connection to other modules in the Vanquish system
6
USB (Universal Serial Bus) port ("B" type connector)
Allows connection to other modules in the Vanquish system or the computer on which the data management system is installed, such as the Chromeleon software
7
Rating plate, indicating the serial number, part number, module name, revision number (if any), and the line and fuse rating
8
Gas inlet
Allows connection to a gas supply
9
Gas exhaust
Allows connection to ventilation
TIP
Thermo Fisher Scientific recommends using the USB ports only as described above. If the USB ports are used for any other purpose, Thermo Fisher Scientific cannot ensure proper functionality.
Follow these steps
NOTICE
  • Never use defective communication cables. If you suspect that a cable is defective, replace the cable.
  • To ensure trouble-free operation, use only the cables provided by Thermo Fisher Scientific for connecting the device.
  1. Place the detector in the system as required by the system configuration. See System Arrangement.
  2. Connect the required interface cables to the detector. See Connecting the Interface Cables.
  3. Connect the power cord (see Connecting the Power Cord).