CAUTION
Possible Contamination
Hazardous substances may have contaminated the device during operation and may cause personal injury to service personnel.
  • Decontaminate all parts of the device that you want to return for repair.
  • Fill in and sign the Health and Safety Form. Thermo Fisher Scientific refuses to accept devices for repair if the Health and Safety Form is missing, incompletely filled in, or unsigned.
Preparations
  1. Remove the slide-in module from the enclosure. See Removing the Slide-In Module.
Follow these steps
  1. Install the slide-in module to the enclosure of the replacement device. Follow the steps for inserting the slide-in module in the enclosure and tightening the captive screws in Installing the Slide-In Module.
  1. Follow the instructions in Shipping the Device.
NOTICE
Shipping the slide-in module improperly leads to damage to the device.
Always ship the slide-in module as described in this operating manual.